Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111286 Supplier Invitrogen™ Supplier No. PA5111286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTTVMQGFNR SERCPRDTRI VQLVF Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P2RY13
Gene Accession No. Q9BPV8
Gene Alias 2010001L06Rik; FKSG77; G protein-coupled receptor 68; G protein-coupled receptor 86; GPCR1; GPR86; GPR94; G-protein coupled receptor 86; G-protein coupled receptor 94; P2ry13; P2Y purinoceptor 13; P2Y13; P2yr13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13; purinergic receptor P2Y13; SP174; Vpurinergic receptor P2Y, G-protein coupled 13
Gene Symbols P2RY13
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human P2Y13. Recombinant protein control fragment (Product #RP-95328).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.