Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2Y14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534088
Description
Percent identity with other species by BLAST analysis: Human (100%) Monkey, Marmoset, Mouse (94%) Rat, Hamster, Panda, Dog, Bat, Rabbit, Pig (88%).
GPR105 has been reported to be expressed in brain. ESTs have been isolated from kidney, placenta, uterus, and heart/melanocyte/uterus libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
P2Y14 | |
Polyclonal | |
Unconjugated | |
P2RY14 | |
A330108O13Rik; BPR105; G protein coupled receptor for UDP-glucose; G protein-coupled receptor 105; G protein-coupled receptor VTR 15-20; GPCR105; Gpr105; G-protein coupled receptor 105; KIAA0001; P2ry14; P2Y purinoceptor 14; P2Y(14) receptor; P2Y14; P2Y14 receptor; purinergic receptor P2Y, G-protein coupled, 14; purinergic receptor P2Y14; UDP-glucose receptor; VTR 15-20; VTR 15-20 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q15391 | |
P2RY14 | |
Synthetic 16 amino acid peptide from 2nd extracellular domain of human P2RY14 / P2Y14. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction