Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530113 Supplier Invitrogen™ Supplier No. PA530113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain, rat brain. Predicted reactivity: Human (99%), Mouse (84%), Rat (85%), Pig (82%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to both adenosine and uridine nucleotides. It may participate in control of the cell cycle of endometrial carcinoma cells. Three transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y2
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7
Gene P2RY2
Gene Accession No. P41231
Gene Alias ATP receptor; HP2U; P2RU1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12
Gene Symbols P2RY2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human P2Y2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5029
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.