Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985954
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5954 Supplier Bioss Supplier No. BS4180R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to both adenosine and uridine nucleotides. It may participate in control of the cell cycle of endometrial carcinoma cells. Three transcript variants encoding the same protein have been identified for this gene.

Specifications

Antigen P2Y2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene P2RY2
Gene Accession No. P41231
Gene Alias ATP receptor; HP2U; P2ru1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12
Gene Symbols P2RY2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human P2Y2R.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5029
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.