Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y4 Monoclonal Antibody (3B4H6)
GREENER_CHOICE

Catalog No. PIMA531823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531823 Supplier Invitrogen™ Supplier No. MA531823
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Amino acid sequence of the immunogen is proprietary. This antibody has been tested in indirect ELISA.

Receptor for UTP and UDP coupled to G-proteins that activate a phosphatidylinositol-calcium second messenger system. Not activated by ATP or ADP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3B4H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene P2RY4
Gene Accession No. P51582
Gene Alias NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor
Gene Symbols P2RY4
Host Species Mouse
Immunogen Purified recombinant fragment of human P2RY4 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.