Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y4/P2RY4 Antibody (3B4C11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | P2Y4/P2RY4 |
---|---|
Clone | 3B4C11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61759
![]() |
Novus Biologicals
NBP261759 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
P2Y4/P2RY4 Monoclonal specifically detects P2Y4/P2RY4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
P2Y4/P2RY4 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
P2P, P2Y4P2Y purinoceptor 4, pyrimidinergic receptor P2Y, G-protein coupled, 4, UNRNRUUridine nucleotide receptor | |
P2RY4 | |
IgG1 | |
Protein G purified |
3B4C11 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Purified | |
GPCR | |
5030 | |
Purified recombinant fragment of human P2RY4 (AA: extra mix) expressed in E. Coli. | |
Primary | |
41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title