Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y4/P2RY4 Antibody (3B4H6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261760
Description
P2Y4/P2RY4 Monoclonal specifically detects P2Y4/P2RY4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
P2Y4/P2RY4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:200, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
P2P, P2Y4P2Y purinoceptor 4, pyrimidinergic receptor P2Y, G-protein coupled, 4, UNRNRUUridine nucleotide receptor | |
Mouse | |
41 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3B4H6 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
P2RY4 | |
Purified recombinant fragment of human P2RY4 (AA: extra mix) expressed in E. Coli. | |
Protein G purified | |
GPCR | |
5030 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction