Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y4 Polyclonal Antibody
P2Y4 Polyclonal Antibody
Supplier: Thermo Scientific PA550523
Description
P2Y4 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
P2Y4 | |
Polyclonal | |
Unconjugated | |
P2RY4 | |
NRU; P2P; P2RY4; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; P2Y4R; P2YR4; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y4; RP13-26D14.5; UNR; Uridine nucleotide receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5030 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P51582 | |
P2RY4 | |
Synthetic peptide corresponding to residues near the C terminal of human pyrimidinergic receptor P2Y, G-protein coupled, 4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction