Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2Y6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109850
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109850 Supplier Invitrogen™ Supplier No. PA5109850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFLLPFAALL ACYCLLACRL CRQDGPAEPV AQERRGKAAR MAVVVAAAFA ISFLPFHITK TAYLAVRSTP GVPCTVLEAF AAAYKGTRPF ASANSVLDPI L.

P2Y6 belongs to the family of G protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. P2Y6 expression has been documented in various blood cells, bone marrow, brain, colon, heart, kidney, ovary, placenta, spleen and thymus. ESTs have been isolated from cervix, embryo, eye, kidney, liver/spleen, placenta, and vessel libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene P2ry6
Gene Accession No. Q15077, Q9ERK9
Gene Alias 2010204J23Rik; G-coupled nucleotide receptor; MGC15335; P2 purinoceptor; P2ry6; P2Y ATP receptor 6; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; P2Y6 receptor; PP2891; Purinoceptor p2y6; pyrimidinergic receptor P2Y, G-protein coupled, 6; pyrimidinergic receptor P2Y6
Gene Symbols P2ry6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Purinergic Receptor P2Y6 (NP_7897661).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233571, 5031
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.