Learn More
Invitrogen™ P2Y8 Polyclonal Antibody

Description
Immunogen sequence: GKSYYHVYKL TLCLSCLNNC LDPFVYYFAS REFQLRLREY LGCRRVPRDT LDTRRESLFS ARTTSVRSEA GAHPEGMEGA TRPGLQRQES VF Highest antigen sequence indentity to the following orthologs: Rat - 29%, Mouse - 28%.
Specifications
Specifications
| Antigen | P2Y8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | P2RY8 |
| Gene Accession No. | Q86VZ1 |
| Gene Alias | G-protein coupled purinergic receptor P2Y8; MGC50878; P2RY8; P2Y purinoceptor 8; P2Y8; purinergic receptor P2Y, G-protein coupled, 8; purinergic receptor P2Y8; RP11-261P4.4 |
| Gene Symbols | P2RY8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.