Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p34-Arc/ARPC2 Rabbit anti-Human,Bovine,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™

Catalog No. 07227I100MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07227I100MI 100 μg
07227I25UMI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07227I100MI Supplier MilliporeSigma Supplier No. 07227I100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p34-Arc/ARPC2 Polyclonal antibody specifically detects p34-Arc/ARPC2 in Human,Bovine,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p34-Arc/ARPC2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate KLH
Gene Accession No. NP_005722
Gene Alias Actin-related protein 2/3 complex subunit 2;Arp2/3 complex 34 kDa subunit;p34-ARC
Gene Symbols ARPC2, ARC34, PRO2446
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Actin-related protein 2/3 complex subunit 2 (p34-ARC).
Molecular Weight of Antigen ∼34 kDa observed; 34.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Anti-Anti-p34-Arc\ARPC2 is a rabbit polyclonal antibody that detects Actin-related protein 2/3 complex subunit 2. It targets an epitope with in 14 amino acids from the C-terminal region.
Target Species Human, Bovine, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.