Learn More
p35 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | p35 |
|---|---|
| Concentration | 0.96 mg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2298
![]()
|
Rockland Immunochemicals
600401ME9 |
100 μg |
Each for $661.74
|
|
|||||
50-199-2299
![]()
|
Rockland Immunochemicals
600401ME9S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse p35/Cdk5. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to rat,human, bovine, and Spermophilus citellus.
The protein encoded by this gene (p35) is a neuron-specific activator of cyclin-dependent kinase 5 (CDK5); the activation of CDK5 is required for proper development of the central nervous system. The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form. The cleavage of p35 into p25 results in relocalization of the protein from the cell periphery to nuclear and perinuclear regions. P25 deregulates CDK5 activity by prolonging its activation and changing its cellular location. The p25 form accumulates in the brain neurons of patients with Alzheimer's disease. This accumulation correlates with an increase in CDK5 kinase activity, and may lead to aberrantly phosphorylated forms of the microtubule-associated protein tau, which contributes to Alzheimer's disease.Specifications
| p35 | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P61809, Q15078 | |
| 12569, 8851 | |
| Primary | |
| Antibody |
| 0.96 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| CDK5 activator 1; CDK5P35; Cdk5r; Cdk5r1; cyclin dependent kinase 5 regulatory subunit 1; Cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 activator 1, p25; Cyclin-dependent kinase 5 activator 1, p35; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); D11Bwg0379e; Nck5a; neuronal CDK5 activator; p23; p25; p35; p35nck5a; regulatory partner for CDK5 kinase; Tau protein kinase II 23 kDa subunit; tau protein kinase II 23kDa subunit; TPKII regulatory subunit | |
| Cdk5r1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Cdk5r1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.