Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 alpha Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11096907UV
Description
p38 alpha Polyclonal antibody specifically detects p38 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
p38 alpha | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A | |
A 20 residue synthetic peptide based on the human p38 with the cysteine residue added and coupled to KLH | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
1432 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction