Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 alpha Antibody (M548), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323234
Description
p38 alpha Monoclonal antibody specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
p38 alpha | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
M548 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50 | |
CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A | |
Clone M548 was generated from a recombinant protein corresponding to amino acid residues in the C-terminal region of human p38a. This sequence is identical in rat and mouse p38a. [UniProt# Q16539] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
1432 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction