Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323232
Description
p38 alpha Polyclonal antibody specifically detects p38 alpha in Human, Mouse, Rat, Chicken samples. It is validated for Western BlotSpecifications
p38 alpha | |
Polyclonal | |
Western Blot 1:1000 | |
CSBP;CSBP1;CSBP2;CSPB1;EXIP;MAPK14;Mitogen-activated protein kinase;Mitogen-activated protein kinase 14;Mxi2;p38;p38ALPHA;PRKM14;PRKM15;RK;SAPK2A | |
p38a MAP Kinase synthetic peptide (coupled to KLH) corresponding to amino acid residues from the C-terminus of mouse p38a. This peptide sequence is highly conserved in human and rat p38a, and has high homology to the conserved site in p38 beta. [UniProt# P47811] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
1432 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction