Learn More
Promega p38 beta Kinase Enzyme System, 1mg
Catalog No. PRV6454 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6454 1 mg
1 options
Catalog No. PRV6454 Supplier Promega Supplier No. V6454
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human p38 beta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p38 beta is a member of the p38 MAP kinase family and is activated by both proinflammatory cytokines and environmental stress.
  • Easily Screen and Profile p38β Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C