Learn More
Promega p38 delta Kinase Enzyme System, 10microg
Catalog No. PRV4078 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4078 10 μg
1 options
Catalog No. PRV4078 Supplier Promega Supplier No. V4078
Recombinant full-length human p38 delta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. It is a member of the p38 MAPK family and is activated by chemical and environmental stresses as well as by proinflammatory cytokines.
  • Easily Screen and Profile p38δ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C