Learn More
Promega p38 gamma Kinase Enzyme System, 1mg
Catalog No. PRV3372 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3372 1 mg
1 options
Catalog No. PRV3372 Supplier Promega Supplier No. V3372
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human p38 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p38 gamma is a member of the p38 MAPK family, which is activated in response to stress.
  • Easily Screen and Profile p38γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C