Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 gamma/SAPK3 Antibody (OTI10E1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273189PE
Description
p38 gamma/SAPK3 Monoclonal specifically detects p38 gamma/SAPK3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
p38 gamma/SAPK3 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
MAPK12 | |
Full-length protein expressed in 293T cell transfected with human p38 gamma/SAPK3 expression vector. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI10E1 | |
Flow Cytometry | |
EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
6300.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction