Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p38 MAPK beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581217 Supplier Invitrogen™ Supplier No. PA581217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MAPK11.

MAPK11 (p38 beta) is a serine/threonine kinase which is involved in a variety of cellular processes, and is activated by proinflammatory cytokines and environmental stress. Mitogen-activated protein kinase 11. The protein encoded by p38 MAPK beta is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. This kinase is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 MAPK beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MAPK11
Gene Accession No. Q15759
Gene Alias MAP kinase 11; MAP kinase p38 beta; MAPK 11; Mapk11; mitogen activated protein kinase 11; mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; OTTHUMP00000196655; p38-2; P38b; p38beta; P38BETA2; Prkm11; protein kinase, mitogen activated kinase, 11, p38beta; SAPK2; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Symbols MAPK11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human MAPK11.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5600
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.