Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p38 MAPK gamma Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580792
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ERK3/MAPK12/P38-gamma.
MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes. It is activated during hypoxia and gamma-irradiation-induced G(2) arrest. Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.
Specifications
p38 MAPK gamma | |
Polyclonal | |
Unconjugated | |
Mapk12 | |
AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
29857, 60352, 6300 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
O08911, P53778, Q63538 | |
Mapk12 | |
Recombinant Human ERK3/MAPK12/P38-gamma Protein (Met1-Leu367). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction