Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144483 Supplier Invitrogen™ Supplier No. PA5144483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p39 Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this gene is a neuron-specific activator of CDK5 kinase. It associates with CDK5 to form an active kinase. This protein and neuron-specific CDK5 activator CDK5R1/p39NCK5A both share limited similarity to cyclins, and thus may define a distinct family of cyclin-dependent kinase activating proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p39
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene 39
Gene Accession No. Q9T1Q9
Gene Alias B230310J22Rik; CDK5 activator 2; Cdk5r2; cyclin dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5 activator 2; cyclin-dependent kinase 5 activator isoform p39i; cyclin-dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5, regulatory subunit 2 (p39); NCK5AI; neuronal CDK5 activator isoform; p39; p39I; p39nck5ai; RGD1562530
Gene Symbols 39
Host Species Rabbit
Immunogen Recombinant Acyrthosiphon pisum secondary endosymbiont phage 1 (Bacteriophage APSE-1) VP39 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1262333
Target Species Insect
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.