Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517177
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a neuron-specific activator of CDK5 kinase. It associates with CDK5 to form an active kinase. This protein and neuron-specific CDK5 activator CDK5R1/p39NCK5A both share limited similarity to cyclins, and thus may define a distinct family of cyclin-dependent kinase activating proteins.
Specifications
| p39 | |
| Polyclonal | |
| Unconjugated | |
| CDK5R2 | |
| B230310J22Rik; CDK5 activator 2; Cdk5r2; cyclin dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5 activator 2; cyclin-dependent kinase 5 activator isoform p39i; cyclin-dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5, regulatory subunit 2 (p39); NCK5AI; neuronal CDK5 activator isoform; p39; p39I; p39nck5ai; RGD1562530 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12570, 501164, 8941 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 20 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O35926, Q13319 | |
| CDK5R2 | |
| Synthetic peptide surrounding Thr90 of human p39. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction