Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P3NP Monoclonal Antibody (C2)

Catalog No. PIMA543877
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543877 20 μL
PIMA541663 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543877 Supplier Invitrogen™ Supplier No. MA543877
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

P3NP Monoclonal Antibody for Western Blot

Procollagen type III N-terminal peptide (P3NP) is released as part of collagen synthesis process in muscle. The increase of circulating P3NP is a marker not only of muscle growth, but also of muscle repair and fibrosis. P3NP may be a potential biomarker for sarcopenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P3NP
Applications Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COL3A1
Gene Accession No. P02461
Gene Alias PIIINP gene; Procollagen 3 N-Terminal Propeptide; Procollagen III N-Terminal Propeptide
Gene Symbols COL3A1
Host Species Mouse
Immunogen Recombinant protein Procollagen III N-Terminal Propeptide.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.