Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p40/deltaNp63 Rabbit anti-Human, Clone: P40/7302R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31380620UG
This item is not returnable.
View return policy
Description
p40/deltaNp63 Monoclonal antibody specifically detects p40/deltaNp63 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
p40/deltaNp63 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Q9H3D4 | |
Rabbit | |
Protein A purified | |
RUO | |
8626 | |
Store at 4°C. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
P40/7302R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
AIS, amplified in squamous cell carcinoma, B(p51A), B(p51B), Chronic ulcerative stomatitis protein, EEC3, Keratinocyte transcription factor KET, KET, LMS, OFC8, p53CP, SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein p53-competing protein | |
Recombinant fragment (around aa1-200) of human p40/deltaNp63 protein (exact sequence is proprietary) (Uniprot: Q9H3D4 ) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction