Learn More
Invitrogen™ p40 Monoclonal Antibody (4C9)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA520977
Catalog No. PIMA520977
Only null left
Add to Cart
Description
Description
Peptide Sequence: YHTEEQHWTL LKFDTLLPPG RLDHSMCIIP WPVTCASEKE DSNSLTLNHE AEKEDSADKV MSHSGDSHEE SQTATLLCLV FGGMNTEGEI YDDCIVTVVD.
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p40
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q7Z6M1
Rabepk
RABEPK (NP_005824.2, 273 a.a. ~ 372 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
4C9
Unconjugated
Rabepk
40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612
Mouse
Affinity chromatography
RUO
10244
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.