Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P40 Monoclonal Antibody (ZY601), Abbomax

Catalog No. 502204416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4416 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4416 Supplier Abbomax Inc Supplier No. 606120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: squamous cell carcinoma Cellular location: Cytoplasm.

P40 Monoclonal antibody specifically detects P40 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen P40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ZY601
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Rabepk
Gene Accession No. Q7Z6M1
Gene Alias 40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612
Host Species Mouse
Immunogen A recombinant protein P40
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.