Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144730 Supplier Invitrogen™ Supplier No. PA5144730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKQLPVLEPG DKPRKATWYT LTVPGDSPCA RVGHSCSYLP PVGNAKRGKV FIVGGANPNR SFSDVHTMDL GKHQWDLDTC KGLLPRYEHA SFIPSCTP Antigen sequence identity: Mouse 83%; Rat 82%.

The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p40
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rabepk
Gene Accession No. Q7Z6M1
Gene Alias 40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612
Gene Symbols Rabepk
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-98.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.