Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582855
Description
Immunogen sequence: NCLQVLNPET RTWTTPEVTS PPPSPRTFHT SSAAIGNQLY VFGGGERGAQ PVQDTKLHVF DANTLTWSQP ETLGNPPSPR H.
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
Specifications
p40 | |
Polyclonal | |
Unconjugated | |
Rabepk | |
40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10244 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6M1 | |
Rabepk | |
Recombinant protein corresponding to Human RABEPK. Recombinant protein control fragment (Product #RP-92977). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction