Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p40 Rabbit anti-Human, Clone: S06-8K2, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | p40 |
---|---|
Clone | S06-8K2 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
p40 Monoclonal antibody specifically detects p40 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
p40 | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
40 kDa Rab9 effector protein, bA65N13.1, p40, Rab9 effector p40, Rab9 effector protein with kelch motifs, RAB9P40DKFZp686P1077, RABEPK | |
Recombinant protein of human p40 (Uniprot#: Q7Z6M1) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S06-8K2 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
10244 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title