Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p40 Rabbit anti-Human, Mouse, Rat, Clone: 10E2A4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | p40 |
---|---|
Clone | 10E2A4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
p40 Monoclonal antibody specifically detects p40 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
p40 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10244 | |
IgG | |
Affinity purified |
10E2A4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
40 kDa Rab9 effector protein, bA65N13.1, p40, Rab9 effector p40, Rab9 effector protein with kelch motifs, RAB9P40DKFZp686P1077, RABEPK | |
Recombinant fusion protein containing a sequence corresponding to amino acids 273-372 of human RAB9p40 (Q7Z6M1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title