Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p40 Recombinant Rabbit Monoclonal Antibody (10E2A4)
GREENER_CHOICE

Catalog No. PIMA537928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537928 Supplier Invitrogen™ Supplier No. MA537928
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: A-549, Mouse testis, Mouse brain, Rat testis Immunogen sequence: YHTEEQHWTL LKFDTLLPPG RLDHSMCIIP WPVTCASEKE DSNSLTLNHE AEKEDSADKV MSHSGDSHEE SQTATLLCLV FGGMNTEGEI YDDCIVTVVD.

The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p40
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 10E2A4
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rabepk
Gene Accession No. Q4V8F4, Q7Z6M1, Q8VCH5
Gene Alias 40 kDa Rab9 effector protein; 8430412M01Rik; 9530020d24rik; AV073337; bA65N13.1; C87311; p40; Rab9 effector p40; rab9 effector protein with kelch motifs; RAB9P40; RABEPK; RGD1310612
Gene Symbols Rabepk
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 273-372 of human RAB9RAB9P40 (Q7Z6M1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10244, 227746, 296649
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.