Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p400 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109685 Supplier Invitrogen™ Supplier No. PA5109685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLAPLPLPSP TSPGFQFSAQ PRRFEHGSPS YIQVTSPLSQ QVQTQSPTQP SPGPGQALQN VRAGAPGPGL GLCSSSPTGG FVDASVLVRQ ISLSPSSGGH FVFQDGSGLT QIAQGAQVQL QHPGTPITVR ERRPSQPHTQ SGGTIHHLGP QSPAAAGGAG LQPLASPSHI TTANLPPQIS SIIQGQLVQQ QQVLQGPPLP RPLGFERTPG VLLPGAGGAA GFGMTSPPPP TSPSRTAVPP G.

Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. May be required for transcriptional activation of E2F1 and MYC target genes during cellular proliferation. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. May regulate ZNF42 transcription activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p400
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EP400
Gene Accession No. Q96L91
Gene Alias 1700020J09Rik; AU023439; CAG repeat protein 32; CAGH32; Domino homolog; E1A binding protein p400; E1A-binding protein p400; EP400; hDomino; KIAA1498; KIAA1818; mDomino; mKIAA1498; P400; p400 kDa SWI2/SNF2-related protein; p400 SWI2/SNF2-related protein; TNRC12; trinucleotide repeat containing 12; trinucleotide repeat-containing gene 12 protein
Gene Symbols EP400
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57634
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.