Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p400 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583942
Description
Immunogen sequence: GTIKTSVTGT SMPTGAVSGN VIVNTIAGVP AATFQSINKR LASPVAPGAL TTPGGSAPAQ VVHTQPPPRA VGSPATATPD LVSMATTQGV RAVTSVTASA VVTTNLTPVQ TPARSLVPQV SQAT.
Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. May be required for transcriptional activation of E2F1 and MYC target genes during cellular proliferation. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. May regulate ZNF42 transcription activity.
Specifications
p400 | |
Polyclonal | |
Unconjugated | |
EP400 | |
1700020J09Rik; AU023439; CAG repeat protein 32; CAGH32; Domino homolog; E1A binding protein p400; E1A-binding protein p400; EP400; hDomino; KIAA1498; KIAA1818; mDomino; mKIAA1498; P400; p400 kDa SWI2/SNF2-related protein; p400 SWI2/SNF2-related protein; TNRC12; trinucleotide repeat containing 12; trinucleotide repeat-containing gene 12 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96L91 | |
EP400 | |
Recombinant protein corresponding to Human EP400. Recombinant protein control fragment (Product #RP-106212). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction