Learn More
Invitrogen™ p41-ARCb Polyclonal Antibody

Description
Recommended positive controls: NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | p41-ARCb |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ARPC1B |
| Gene Accession No. | O15143, Q9WV32 |
| Gene Alias | 41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs |
| Gene Symbols | ARPC1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.