Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p41-ARCb Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518047
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518047 Supplier Invitrogen™ Supplier No. PA518047
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. This protein also has a role in centrosomal homeostasis by being an activator and substrate of the Aurora A kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p41-ARCb
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ARPC1B
Gene Accession No. O15143
Gene Alias 41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs
Gene Symbols ARPC1B
Host Species Goat
Immunogen Synthetic peptide sequence (AYHSFLVEPISCH) corresponding to the N-terminus amino acids of ARPC1B (aa 2-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10095
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.