Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p41 Flagellin Antibody (6802), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308849G
Description
p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
p41 Flagellin | |
Monoclonal | |
DyLight 488 | |
41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41 | |
A crude sonicate of Borrelia spirochetes | |
100 μg | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunofluorescence | |
6802 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
46847386 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction