Learn More
Promega P450-Glo CYP2C19 Screening System, 1,000 assays
Catalog No. PRV9880 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRV9880 1000 Assays
1 options
Catalog No. PRV9880 Supplier Promega Supplier No. V9880
The P450-Glo Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays
  • A Luminescent Method and Screening System for Measuring CYP2C19 Activity
  • Screening system includes complete set of reagents
  • Luminescent format eliminates need for time−consuming analyses
  • Broad dynamic range and low background
Specifications
Quantity 1000 Assays
Content And Storage Components at different temperatures