Learn More
Promega P450-Glo™ CYP3A4 Assay (Luciferin-PPXE) DMSO-Tolerant Assay
Shop All Promega Corporation Products
Description
Includes
Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
- Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
- Screening systems include complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer, 2 x 15μL Luciferin-PPXE <-65°C |
| Detection Method | Homogeneous, Luminescence |
| For Use With (Application) | For measuring cytochrome P450 activity |
| Product Type | P450-Glo CYP3A4 Assay (Luciferin-PPXE) DMSO-Tolerant Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.