Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P4HA1 Polyclonal Antibody
P4HA1 Polyclonal Antibody
Supplier: Thermo Scientific PA531246
Description
P4HA1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
P4HA1 | |
Polyclonal | |
Unconjugated | |
P4HA1 | |
4-PH alpha-1; AL022634; collagen prolyl 4-hydroxylase alpha(I); C-P4Halpha(I); P4ha; P4ha1; PHalphaI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha 1 polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-1; prolyl 4-hydroxylase alpha subunit; prolyl 4-hydroxylase subunit alpha 1; prolyl 4-hydroxylase subunit alpha-1; prolyl 4-hydroxylase, alpha polypeptide I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5033 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P13674 | |
P4HA1 | |
Recombinant fragment corresponding to a region within amino acids 318 and 534 of Human P4HA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction