Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P4HA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552454 Supplier Invitrogen™ Supplier No. PA552454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTSIGQMTDL IHTEKDLVTS LKDYIKAEED KLEQIKKWAE KLDRLTSTAT KDPEGFVGHP VNAFKLMKRL NTEWSELENL VLKDMSDGFI SNLTIQRQYF PNDEDQVGAA KALLRLQDTY NLDTDTISKG NLPGVKHKSF LTAEDC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P4HA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P4HA1
Gene Accession No. P13674
Gene Alias 4-PH alpha-1; AL022634; collagen prolyl 4-hydroxylase alpha(I); C-P4Halpha(I); P4ha; P4ha1; PHalphaI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha 1 polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-1; prolyl 4-hydroxylase alpha subunit; prolyl 4-hydroxylase subunit alpha 1; prolyl 4-hydroxylase subunit alpha-1; prolyl 4-hydroxylase, alpha polypeptide I
Gene Symbols P4HA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human P4HA1. Recombinant protein control fragment (Product #RP-89690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.