Learn More
Invitrogen™ P4HA2 Polyclonal Antibody

Description
Recommended positive controls: H1299, HeLa. Predicted reactivity: Mouse (93%), Rat (93%), Chicken (81%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | P4HA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.82 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.01% thimerosal; pH 7 |
| Gene | P4HA2 |
| Gene Accession No. | O15460 |
| Gene Alias | 4-PH alpha 2; 4-PH alpha-2; 4PHalpha2; AA407196; C76437; collagen prolyl 4-hydroxylase alpha(II); C-P4Halpha(II); hypothetical protein LOC507327; P4HA2; P4hl; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide 2; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; prolyl 4-hydroxylase A; Prolyl 4-hydroxylase alpha IIa subunit; prolyl 4-hydroxylase subunit alpha 2; prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II; UNQ290/PRO330; zgc:91948 |
| Gene Symbols | P4HA2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.