Learn More
Invitrogen™ P4HA3 Monoclonal Antibody (7F10)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524534
Catalog No. PIMA524534
Only null left
Add to Cart
Description
Description
Peptide Sequence: TRFYDKVLSL HEDSTTPVAN PLLAFTLIKR LQSDWRNVVH SLEASENIRA LKDGYEKVEQ DLPAFEDLEG AARALMRLQD VYMLNVKGLA RGVFQRVTGS.
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P4HA3
Monoclonal
Unconjugated
P4ha3
4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374
Mouse
Protein A
RUO
283208
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
7F10
PBS with no preservative; pH 7.4
Q7Z4N8
P4ha3
P4HA3 (NP_878907.1, 66 a.a. ~ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.