Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P4HA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548623
Description
P4HA3 Polyclonal Antibody for Western Blot
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
Specifications
P4HA3 | |
Polyclonal | |
Unconjugated | |
P4ha3 | |
4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
283208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q7Z4N8 | |
P4ha3 | |
KLH conjugated synthetic peptide between 274-303 amino acids from the Central region of human P4HA3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction