Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P4HA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552483 Supplier Invitrogen™ Supplier No. PA552483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTPVANPLLA FTLIKRLQSD WRNVVHSLEA SENIRALKDG YEKVEQDLPA FEDLEGAARA LMRLQDVYML NVKGLARGVF QRVTGSAITD LYSPKRLFSL TGDDCFQVGK V Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P4HA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P4ha3
Gene Accession No. Q7Z4N8
Gene Alias 4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374
Gene Symbols P4ha3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human P4HA3. Recombinant protein control fragment (Product #RP-89693).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.