Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P4HB Rabbit anti-Human, Mouse, Rat, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30179
Description
Rabbit polyclonal antibody raised against partial synthetic peptide of human P4HB.
- High specificity
- High reactivity
Specifications
P4HB | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic peptide of human P4HB. | |
Lyophilized from 0.5 mg/mL PBS, pH 7.4 (0.02% sodium azide). | |
DSI/ERBA2L/GIT/P4Hbeta/PDI/PDIA1/PHDB/PO4DB/PO4HB/PROHB | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
5034 | |
Store at -20°C or lower. Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. The reconstituted antibody can be stored for 2-3 weeks at 4°C. For long term storage, aliquot and st |
|
IgG |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
ELISA (0.05-0.2 ug/mL) Flow Cytometry (1-3 ug/106 cells) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P07237 | |
P4HB | |
A synthetic peptide (conjugated with KLH) corresponding to residues within 100-150 of human P4HB. | |
40 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction