Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p50 dynamitin Polyclonal Antibody, Huabio

Catalog No. 502544626
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4626 Supplier HUABIO RESEARCH INC Supplier No. HA500029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p50 dynamitin Polyclonal antibody specifically detects p50 dynamitin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen p50 dynamitin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene DCTN2
Gene Accession No. Q13561, Q6AYH5, Q99KJ8
Gene Alias 2310042E05Rik; 50 kDa dynein-associated polypeptide; C130077D06Rik; C87049; DCTN2; dctn2 protein; DCTN50; DCTN-50; dynactin 2; dynactin 2 (p50); dynactin complex 50 kDa subunit; Dynactin subunit 2; dynamitin; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; HEL-S-77; hypothetical protein LOC527201; p50; p50 dynamitin; p50/dynamitin; RBP50; zgc:63867
Host Species Rabbit
Immunogen Recombinant protein within human p50 dynamitin aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10540, 299850, 69654
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.