Learn More
Description
Host Species: Mouse
Clone: DO-7 + BP53-12
Isotype: IgG2b + IgG2a
Species Reactivity: Human. Others not known.
Epitope: aa 37-45 and aa 20-25
Immunogen: Recombinant human p53 protein
Molecular Weight: 53kDa
Positive Control: MDA-231, SKBR-3 cells or Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Specifications
Specifications
| Antigen | p53 Ab-8 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | DO-7 + BP53-12 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human p53 protein |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.