Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 Antibody (Pab DO-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | p53 |
---|---|
Clone | Pab DO-1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay |
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)Specifications
p53 | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
PBS | |
7157 | |
IgG2a | |
Protein A purified |
Pab DO-1 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
Recombinant human wild type p53 protein expressed in E.coli | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title