Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p53 Antibody (PAb421) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 3 publications
$281.00
Specifications
Antigen | p53 |
---|---|
Clone | PAb421 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, In vivo assay, Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
p53 Monoclonal specifically detects p53 in Human, Mouse, Rat, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, Immunofluorescence.Specifications
p53 | |
Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, In vivo assay, Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Rabbit | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
TP53 | |
IgG κ | |
Recognises the mammalian mutant and wild-type p53 protein and reacts with an epitope between amino acid residues 370 and 378 (near the C-terminus of the protein). |
PAb421 | |
Monoclonal | |
Purified | |
PBS | |
7157 | |
Synthetic peptide corresponding to aa 371-380 of human p53. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title